Product Details

SNP ID
rs201781094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32442666 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCAACCTGGAAATCATGACAAAG[A/C]GCTCCAACTATACTCCGATCACCAA
Phenotype
MIM: 142860
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HLA-DRA PubMed Links

Gene Details

Gene
HLA-DRA
Gene Name
major histocompatibility complex, class II, DR alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019111.4 410 Missense Mutation AGC,CGC S101R NP_061984.2

View Full Product Details