Product Details

SNP ID
rs201978922
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:109444746 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACCTTCCCGGTTCATGTAGCCTC[A/G]TAGCTCCTGGTCCAGCTGCCACTGT
Phenotype
MIM: 607129 MIM: 603498
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MICAL1 PubMed Links

Gene Details

Gene
MICAL1
Gene Name
microtubule associated monooxygenase, calponin and LIM domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159291.1 3382 Nonsense Mutation CGA,TGA R926* NP_001152763.1
NM_001286613.1 3382 Nonsense Mutation CGA,TGA R1031* NP_001273542.1
NM_022765.3 3382 Nonsense Mutation CGA,TGA R1012* NP_073602.3
Gene
PPIL6
Gene Name
peptidylprolyl isomerase like 6
There are no transcripts associated with this gene.

Gene
SMPD2
Gene Name
sphingomyelin phosphodiesterase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003080.2 3382 Intron NP_003071.2
XM_005267109.1 3382 Intron XP_005267166.1
XM_011536079.1 3382 Intron XP_011534381.1

View Full Product Details