Product Details

SNP ID
rs200403445
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41737813 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCTGGGCAGATTCTGAATGCTGT[G/T]ACAGTTCACGAGAAACTGCAAGAGG
Phenotype
MIM: 169740 MIM: 600744
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PGC PubMed Links

Gene Details

Gene
PGC
Gene Name
progastricsin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166424.1 998 Intron NP_001159896.1
NM_002630.3 998 Missense Mutation AAC,CAC N311H NP_002621.1
Gene
TFEB
Gene Name
transcription factor EB
There are no transcripts associated with this gene.

View Full Product Details