Product Details
- SNP ID
-
rs201293893
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:56463579 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGAGTCTTACCTTGGGTGTGGTGG[G/T]GGCAGGGACCTGTGGGGAGGCCGCC
- Phenotype
-
MIM: 113810
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DST
PubMed Links
Gene Details
- Gene
- DST
- Gene Name
- dystonin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001144769.2 |
16729 |
Missense Mutation |
ACC,CCC |
T5506P |
NP_001138241.1 |
NM_001144770.1 |
16729 |
Missense Mutation |
ACC,CCC |
T5368P |
NP_001138242.1 |
NM_001723.5 |
16729 |
Intron |
|
|
NP_001714.1 |
NM_015548.4 |
16729 |
Missense Mutation |
ACC,CCC |
T5002P |
NP_056363.2 |
NM_183380.3 |
16729 |
Missense Mutation |
ACC,CCC |
T5328P |
NP_899236.1 |
XM_005249310.3 |
16729 |
Missense Mutation |
ACC,CCC |
T7649P |
XP_005249367.1 |
XM_005249315.3 |
16729 |
Missense Mutation |
ACC,CCC |
T7616P |
XP_005249372.1 |
XM_005249316.2 |
16729 |
Missense Mutation |
ACC,CCC |
T7554P |
XP_005249373.1 |
XM_005249318.2 |
16729 |
Missense Mutation |
ACC,CCC |
T7511P |
XP_005249375.1 |
XM_005249319.2 |
16729 |
Missense Mutation |
ACC,CCC |
T7478P |
XP_005249376.1 |
XM_005249320.4 |
16729 |
Missense Mutation |
ACC,CCC |
T7471P |
XP_005249377.1 |
XM_005249322.4 |
16729 |
Missense Mutation |
ACC,CCC |
T7464P |
XP_005249379.1 |
XM_005249323.3 |
16729 |
Missense Mutation |
ACC,CCC |
T7438P |
XP_005249380.1 |
XM_005249324.2 |
16729 |
Missense Mutation |
ACC,CCC |
T7112P |
XP_005249381.1 |
XM_011514824.2 |
16729 |
Missense Mutation |
ACC,CCC |
T7625P |
XP_011513126.1 |
XM_011514825.2 |
16729 |
Missense Mutation |
ACC,CCC |
T5515P |
XP_011513127.1 |
XM_011514826.2 |
16729 |
Missense Mutation |
ACC,CCC |
T7329P |
XP_011513128.1 |
XM_017011205.1 |
16729 |
Missense Mutation |
ACC,CCC |
T7658P |
XP_016866694.1 |
XM_017011206.1 |
16729 |
Missense Mutation |
ACC,CCC |
T7652P |
XP_016866695.1 |
XM_017011207.1 |
16729 |
Missense Mutation |
ACC,CCC |
T7637P |
XP_016866696.1 |
XM_017011208.1 |
16729 |
Missense Mutation |
ACC,CCC |
T7634P |
XP_016866697.1 |
XM_017011209.1 |
16729 |
Missense Mutation |
ACC,CCC |
T7628P |
XP_016866698.1 |
XM_017011210.1 |
16729 |
Missense Mutation |
ACC,CCC |
T7658P |
XP_016866699.1 |
XM_017011211.1 |
16729 |
Missense Mutation |
ACC,CCC |
T7634P |
XP_016866700.1 |
XM_017011212.1 |
16729 |
Missense Mutation |
ACC,CCC |
T7580P |
XP_016866701.1 |
XM_017011213.1 |
16729 |
Missense Mutation |
ACC,CCC |
T7549P |
XP_016866702.1 |
XM_017011214.1 |
16729 |
Missense Mutation |
ACC,CCC |
T7525P |
XP_016866703.1 |
XM_017011215.1 |
16729 |
Missense Mutation |
ACC,CCC |
T7549P |
XP_016866704.1 |
XM_017011216.1 |
16729 |
Missense Mutation |
ACC,CCC |
T7525P |
XP_016866705.1 |
XM_017011217.1 |
16729 |
Missense Mutation |
ACC,CCC |
T7450P |
XP_016866706.1 |
XM_017011218.1 |
16729 |
Missense Mutation |
ACC,CCC |
T5847P |
XP_016866707.1 |
XM_017011219.1 |
16729 |
Missense Mutation |
ACC,CCC |
T5572P |
XP_016866708.1 |
XM_017011220.1 |
16729 |
Missense Mutation |
ACC,CCC |
T5530P |
XP_016866709.1 |
XM_017011221.1 |
16729 |
Missense Mutation |
ACC,CCC |
T5494P |
XP_016866710.1 |
XM_017011222.1 |
16729 |
Missense Mutation |
ACC,CCC |
T5468P |
XP_016866711.1 |
XM_017011223.1 |
16729 |
Missense Mutation |
ACC,CCC |
T5463P |
XP_016866712.1 |
XM_017011224.1 |
16729 |
Missense Mutation |
ACC,CCC |
T5243P |
XP_016866713.1 |
XM_017011225.1 |
16729 |
Intron |
|
|
XP_016866714.1 |
View Full Product Details