Product Details

SNP ID
rs200862397
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:72622521 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCGCAACGTCAAGTACCGGCGGG[G/T]GCAGAACTACCTGTACAACGTGCTG
Phenotype
MIM: 607357
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KCNQ5 PubMed Links

Gene Details

Gene
KCNQ5
Gene Name
potassium voltage-gated channel subfamily Q member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160130.1 679 Missense Mutation GGG,GTG G111V NP_001153602.1
NM_001160132.1 679 Missense Mutation GGG,GTG G111V NP_001153604.1
NM_001160133.1 679 Missense Mutation GGG,GTG G111V NP_001153605.1
NM_001160134.1 679 Missense Mutation GGG,GTG G111V NP_001153606.1
NM_019842.3 679 Missense Mutation GGG,GTG G111V NP_062816.2
XM_017011058.1 679 Missense Mutation GGG,GTG G111V XP_016866547.1
Gene
KCNQ5-IT1
Gene Name
KCNQ5 intronic transcript 1
There are no transcripts associated with this gene.

View Full Product Details