Product Details

SNP ID
rs201176928
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:8413550 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTTAAATAGGACAATCACTGTC[A/T]ATACAGTCTGTGCCAGCGTCCTTGA
Phenotype
MIM: 610845
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC35B3 PubMed Links

Gene Details

Gene
SLC35B3
Gene Name
solute carrier family 35 member B3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011514650.2 Intron XP_011512952.1

View Full Product Details