Product Details
- SNP ID
-
rs201335180
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:44229656 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCACTGCTGAACTGAGCAAGGACG[C/T]CCAGGCGTCAGCCGCCCCTGCAGCA
- Phenotype
-
MIM: 602193
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SLC29A1
PubMed Links
Gene Details
- Gene
- SLC29A1
- Gene Name
- solute carrier family 29 member 1 (Augustine blood group)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001078175.2 |
475 |
Missense Mutation |
GCC,GTC |
A60V |
NP_001071643.1 |
NM_001078177.1 |
475 |
Missense Mutation |
GCC,GTC |
A60V |
NP_001071645.1 |
NM_001304462.1 |
475 |
Missense Mutation |
GCC,GTC |
A139V |
NP_001291391.1 |
NM_001304463.1 |
475 |
Missense Mutation |
GCC,GTC |
A102V |
NP_001291392.1 |
NM_001304465.1 |
475 |
Missense Mutation |
GCC,GTC |
A86V |
NP_001291394.1 |
NM_001304466.1 |
475 |
Missense Mutation |
GCC,GTC |
A85V |
NP_001291395.1 |
XM_005248876.4 |
475 |
Missense Mutation |
GCC,GTC |
A103V |
XP_005248933.1 |
XM_005248878.3 |
475 |
Missense Mutation |
GCC,GTC |
A60V |
XP_005248935.1 |
XM_005248879.3 |
475 |
Missense Mutation |
GCC,GTC |
A60V |
XP_005248936.1 |
XM_005248880.3 |
475 |
Missense Mutation |
GCC,GTC |
A60V |
XP_005248937.1 |
XM_005248881.3 |
475 |
Missense Mutation |
GCC,GTC |
A60V |
XP_005248938.1 |
XM_005248882.3 |
475 |
Missense Mutation |
GCC,GTC |
A60V |
XP_005248939.1 |
XM_011514341.2 |
475 |
Missense Mutation |
GCC,GTC |
A140V |
XP_011512643.1 |
View Full Product Details