Product Details

SNP ID
rs200369406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43184324 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGAGGTCAGGATGGTGGGGGAAC[A/G]GCATGCTGGGGACCTCATGGTGCCC
Phenotype
MIM: 607489 MIM: 600589
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CUL9 PubMed Links

Gene Details

Gene
CUL9
Gene Name
cullin 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015089.3 809 Missense Mutation CAG,CGG Q5R NP_055904.1
XM_011514422.1 809 Missense Mutation CAG,CGG Q5R XP_011512724.1
XM_011514423.2 809 Missense Mutation CAG,CGG Q5R XP_011512725.1
XM_011514424.1 809 Missense Mutation CAG,CGG Q5R XP_011512726.1
XM_011514425.1 809 Missense Mutation CAG,CGG Q5R XP_011512727.1
XM_011514426.1 809 Missense Mutation CAG,CGG Q5R XP_011512728.1
XM_011514427.1 809 Intron XP_011512729.1
XM_011514428.1 809 Missense Mutation CAG,CGG Q5R XP_011512730.1
XM_011514429.1 809 Intron XP_011512731.1
XM_011514430.1 809 Missense Mutation CAG,CGG Q5R XP_011512732.1
XM_017010589.1 809 Missense Mutation CAG,CGG Q5R XP_016866078.1
XM_017010590.1 809 Intron XP_016866079.1
Gene
SRF
Gene Name
serum response factor
There are no transcripts associated with this gene.

View Full Product Details