Product Details

SNP ID
rs200548546
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:13615379 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCATGGTGCAGCTCCGACCGCG[A/C]GCGTCTCGCGCCCCGGCGTCGGCGG
Phenotype
MIM: 611533 MIM: 603854 MIM: 604483
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NOL7 PubMed Links

Gene Details

Gene
NOL7
Gene Name
nucleolar protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317724.1 53 Silent Mutation CGA,CGC R7R NP_001304653.1
NM_016167.4 53 Silent Mutation CGA,CGC R7R NP_057251.2
Gene
RANBP9
Gene Name
RAN binding protein 9
There are no transcripts associated with this gene.

Gene
SIRT5
Gene Name
sirtuin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193267.2 53 Intron NP_001180196.1
NM_001242827.1 53 Intron NP_001229756.1
NM_012241.4 53 Intron NP_036373.1
NM_031244.3 53 Intron NP_112534.1
XM_005248968.4 53 Intron XP_005249025.1
XM_011514444.2 53 Intron XP_011512746.1
XM_017010620.1 53 Intron XP_016866109.1
XM_017010621.1 53 Intron XP_016866110.1
XM_017010622.1 53 Intron XP_016866111.1
XM_017010623.1 53 Intron XP_016866112.1
XM_017010624.1 53 Intron XP_016866113.1
XM_017010625.1 53 Intron XP_016866114.1

View Full Product Details