Product Details

SNP ID
rs200650470
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:89894663 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGCCAGGTTGCAACAATATCTG[G/T]TATGATGATGCATTCCCTATCTCTT
Phenotype
MIM: 611924
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GJA10 PubMed Links

Gene Details

Gene
GJA10
Gene Name
gap junction protein alpha 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032602.1 195 Missense Mutation TGG,TGT W65C NP_115991.1

View Full Product Details