Product Details
- SNP ID
-
rs200830616
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:2623588 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGCTCCAGGCCTGACAGCAACCGC[A/G]GTGGGGAGATGGGGCAGGAACTGAG
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LINC01600
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs6902511] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LINC01600
- Gene Name
- long intergenic non-protein coding RNA 1600
There are no transcripts associated with this gene.
View Full Product Details