Product Details

SNP ID
rs201500909
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:75243747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTCCGCAGCATCTTGGCTGTTAC[C/T]GACCAGCAACCGCCACAACTGAACA
Phenotype
MIM: 123996 MIM: 611028
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COX7A2 PubMed Links

Gene Details

Gene
COX7A2
Gene Name
cytochrome c oxidase subunit 7A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001865.3 182 Silent Mutation TCA,TCG S28S NP_001856.2
Gene
TMEM30A
Gene Name
transmembrane protein 30A
There are no transcripts associated with this gene.

View Full Product Details