Product Details

SNP ID
rs200513147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:1394703 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCTTTCTTCTTTCAGTGGGACT[A/G]CCCCGTTACCAGCATCACTCTACTC
Phenotype
MIM: 603250
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FOXF2 PubMed Links

Gene Details

Gene
FOXF2
Gene Name
forkhead box F2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001452.1 1293 Silent Mutation CTA,CTG L393L NP_001443.1
Gene
LOC105374880
Gene Name
uncharacterized LOC105374880
There are no transcripts associated with this gene.

Gene
MIR6720
Gene Name
microRNA 6720
There are no transcripts associated with this gene.

View Full Product Details