Product Details
- SNP ID
-
rs200613112
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:31615536 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACCCCACAGGAGGAAAAGCTTTCG[A/G]ACTGCTGAAGGCCCAGCAGGAAGAG
- Phenotype
-
MIM: 601833
MIM: 142580
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AIF1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2736182] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AIF1
- Gene Name
- allograft inflammatory factor 1
- Gene
- PRRC2A
- Gene Name
- proline rich coiled-coil 2A
There are no transcripts associated with this gene.
- Gene
- SNORA38
- Gene Name
- small nucleolar RNA, H/ACA box 38
There are no transcripts associated with this gene.
View Full Product Details