Product Details

SNP ID
rs200673328
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:50715308 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCACCAGTCCTTCCATTACGAG[A/T]TTCAGCACAGCCACCCGGCCGTCAC
Phenotype
MIM: 610161
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TFAP2D PubMed Links

Gene Details

Gene
TFAP2D
Gene Name
transcription factor AP-2 delta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172238.3 744 Missense Mutation ATT,TTT I78F NP_758438.2

View Full Product Details