Product Details

SNP ID
rs200891668
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:44259225 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCAGACCCTGCCTGGCTTCAGTC[A/G]GTACACAGCAGCAGTTTCCCTGAGA
Phenotype
MIM: 140572 MIM: 604548 MIM: 610788
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
HSP90AB1 PubMed Links

Gene Details

Gene
HSP90AB1
Gene Name
heat shock protein 90 alpha family class B member 1
There are no transcripts associated with this gene.

Gene
MIR4647
Gene Name
microRNA 4647
There are no transcripts associated with this gene.

Gene
NFKBIE
Gene Name
NFKB inhibitor epsilon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004556.2 1522 Silent Mutation NP_004547.2
Gene
SLC35B2
Gene Name
solute carrier family 35 member B2
There are no transcripts associated with this gene.

View Full Product Details