Product Details

SNP ID
rs200988696
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:89178949 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGTCAGCTGCACCATCATCCTGT[C/T]GGGCTTCTCTCCATTCTCTGGCATG
Phenotype
MIM: 137161
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GABRR1 PubMed Links

Gene Details

Gene
GABRR1
Gene Name
gamma-aminobutyric acid type A receptor rho1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256703.1 1665 Missense Mutation NP_001243632.1
NM_001256704.1 1665 Missense Mutation NP_001243633.1
NM_001267582.1 1665 Missense Mutation NP_001254511.1
NM_002042.4 1665 Missense Mutation NP_002033.2
XM_017010689.1 1665 Missense Mutation XP_016866178.1

View Full Product Details