Product Details

SNP ID
rs201206908
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31996082 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGCTGACGGCTCGTTCCAGGAC[A/C]CCTGTCCAGTGTTAGACAGGAGCAT
Phenotype
MIM: 120810
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C4A PubMed Links

Gene Details

Gene
C4A
Gene Name
complement component 4A (Rodgers blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252204.1 Intron NP_001239133.1
NM_007293.2 Intron NP_009224.2
Gene
CYP21A1P
Gene Name
cytochrome P450 family 21 subfamily A member 1, pseudogene
There are no transcripts associated with this gene.

View Full Product Details