Product Details
- SNP ID
-
rs201485985
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:138908713 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCAAGAACAGCAGCTAAGACACCC[A/G]TCTTTTGTGTATGTTCATCAACTTC
- Phenotype
-
MIM: 614825
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ECT2L
PubMed Links
Gene Details
- Gene
- ECT2L
- Gene Name
- epithelial cell transforming 2 like
There are no transcripts associated with this gene.
- Gene
- REPS1
- Gene Name
- RALBP1 associated Eps domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001128617.2 |
2477 |
Missense Mutation |
ACG,ATG |
T697M |
NP_001122089.1 |
NM_001286611.1 |
2477 |
Missense Mutation |
ACG,ATG |
T724M |
NP_001273540.1 |
NM_001286612.1 |
2477 |
Missense Mutation |
ACG,ATG |
T633M |
NP_001273541.1 |
NM_031922.4 |
2477 |
Missense Mutation |
ACG,ATG |
T723M |
NP_114128.3 |
XM_005267177.3 |
2477 |
Missense Mutation |
ACG,ATG |
T723M |
XP_005267234.1 |
XM_005267178.4 |
2477 |
Missense Mutation |
ACG,ATG |
T724M |
XP_005267235.1 |
XM_005267179.3 |
2477 |
Missense Mutation |
ACG,ATG |
T696M |
XP_005267236.1 |
XM_011536202.2 |
2477 |
Missense Mutation |
ACG,ATG |
T679M |
XP_011534504.1 |
XM_017011387.1 |
2477 |
Missense Mutation |
ACG,ATG |
T722M |
XP_016866876.1 |
XM_017011388.1 |
2477 |
Missense Mutation |
ACG,ATG |
T679M |
XP_016866877.1 |
XM_017011389.1 |
2477 |
Missense Mutation |
ACG,ATG |
T652M |
XP_016866878.1 |
View Full Product Details