Product Details

SNP ID
rs200618059
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:73425251 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAAAATTCATCAAAAACTGGAAA[C/T]TCATTGTTTCTTTCATATTCAATTT
Phenotype
MIM: 606286 MIM: 613973
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDX43 PubMed Links

Gene Details

Gene
DDX43
Gene Name
DEAD-box helicase 43
There are no transcripts associated with this gene.

Gene
MB21D1
Gene Name
Mab-21 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138441.2 1684 Silent Mutation GAA,GAG E515E NP_612450.2
XM_017010232.1 1684 Intron XP_016865721.1

View Full Product Details