Product Details

SNP ID
rs200757538
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:77462515 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGTTTCTTCTTTTCCAGCAGGG[A/C]GTCGGAGACTCGCACTTTGACTTGG
Phenotype
MIM: 182131
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HTR1B PubMed Links

Gene Details

Gene
HTR1B
Gene Name
5-hydroxytryptamine receptor 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000863.2 1508 Missense Mutation NP_000854.1

View Full Product Details