Product Details

SNP ID
rs200877202
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:99921072 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGAAAAGGGTTAATGCTGCTGCT[G/T]GCATAGCTGAGACAGATGGAGAGGT
Phenotype
MIM: 606111
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MCHR2 PubMed Links

Gene Details

Gene
MCHR2
Gene Name
melanin concentrating hormone receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040179.1 1206 Silent Mutation GCA,GCC A297A NP_001035269.1
NM_032503.2 1206 Silent Mutation GCA,GCC A297A NP_115892.2

View Full Product Details