Product Details
- SNP ID
-
rs201152908
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:43055599 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAATGGCTGTGCCATTCACCTTCCG[C/T]AGTAGCACACCACACGTCCCTGGGG
- Phenotype
-
MIM: 609577
MIM: 611822
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CUL7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs9471999] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CUL7
- Gene Name
- cullin 7
There are no transcripts associated with this gene.
- Gene
- KLC4
- Gene Name
- kinesin light chain 4
There are no transcripts associated with this gene.
- Gene
- MRPL2
- Gene Name
- mitochondrial ribosomal protein L2
View Full Product Details