Product Details

SNP ID
rs199614275
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:132589129 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCTGGCAACTGCCCACACAAGG[A/C]ATCTCTTCCAGCCACTGGCTGAGCC
Phenotype
MIM: 607405
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TAAR5 PubMed Links

Gene Details

Gene
TAAR5
Gene Name
trace amine associated receptor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003967.2 610 Missense Mutation NP_003958.2

View Full Product Details