Product Details

SNP ID
rs200009734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:132538759 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGGATATGCATTGTTCTTTCCT[A/G]GTTCTTTTCTGTCACATACAGCTTT
Phenotype
MIM: 608282
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TAAR9 PubMed Links

Gene Details

Gene
TAAR9
Gene Name
trace amine associated receptor 9 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175057.3 472 Nonsense Mutation TAG,TGG *157W NP_778227.3

View Full Product Details