Product Details
- SNP ID
-
rs199753878
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:50043647 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTTTGGGATCCAATTATTTGGGTC[C/T]GTAGGGTCACATTCTGTCACGCAGC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB112
PubMed Links
Gene Details
- Gene
- DEFB112
- Gene Name
- defensin beta 112
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001037498.1 |
270 |
Silent Mutation |
ACA,ACG |
T90T |
NP_001032587.1 |
View Full Product Details