Product Details

SNP ID
rs200320516
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:138406042 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTTCAGGTCCTTTTAGTGAGTCT[A/G]CCATTACCATTTCCCTGTATATTCC
Phenotype
MIM: 605825
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HEBP2 PubMed Links

Gene Details

Gene
HEBP2
Gene Name
heme binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001326380.1 607 Missense Mutation ACC,GCC T115A NP_001313309.1
NM_001326381.1 607 Intron NP_001313310.1
NM_014320.2 607 Missense Mutation ACC,GCC T104A NP_055135.1
XM_017010631.1 607 Intron XP_016866120.1

View Full Product Details