Product Details

SNP ID
rs200296788
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:46704631 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTGGTTGTGTTAATGTTGGTCC[C/T]TGGAATAAGATTCTCATCATCTCCT
Phenotype
MIM: 601690 MIM: 611200
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLA2G7 PubMed Links

Gene Details

Gene
PLA2G7
Gene Name
phospholipase A2 group VII
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168357.1 1433 Missense Mutation AGG,GGG R419G NP_001161829.1
NM_005084.3 1433 Missense Mutation AGG,GGG R419G NP_005075.3
XM_005249408.4 1433 Missense Mutation AGG,GGG R419G XP_005249465.1
Gene
TDRD6
Gene Name
tudor domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010870.2 1433 Intron NP_001010870.1
NM_001168359.1 1433 Intron NP_001161831.1

View Full Product Details