Product Details

SNP ID
rs199658474
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:75243725 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGAGAAGCTCCTCACCAGCAGAT[A/T]CCGCAGCATCTTGGCTGTTACTGAC
Phenotype
MIM: 123996 MIM: 611028
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
COX7A2 PubMed Links

Gene Details

Gene
COX7A2
Gene Name
cytochrome c oxidase subunit 7A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001865.3 204 Missense Mutation AAT,TAT N36Y NP_001856.2
Gene
TMEM30A
Gene Name
transmembrane protein 30A
There are no transcripts associated with this gene.

View Full Product Details