Product Details

SNP ID
rs199795809
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41158767 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTCCTCCCATCATCTTCCTTCA[C/T]GTGTCTCTCAGCCCTGCAATAGTCC
Phenotype
MIM: 605086 MIM: 609714
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105375056 PubMed Links

Gene Details

Gene
LOC105375056
Gene Name
uncharacterized LOC105375056
There are no transcripts associated with this gene.

Gene
TREM2
Gene Name
triggering receptor expressed on myeloid cells 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271821.1 794 Missense Mutation ATG,GTG M166V NP_001258750.1
NM_018965.3 794 Silent Mutation ACA,ACG T230T NP_061838.1
XM_006715116.3 794 Silent Mutation ACA,ACG T143T XP_006715179.1
Gene
TREML1
Gene Name
triggering receptor expressed on myeloid cells like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271807.1 794 Intron NP_001258736.1
NM_001271808.1 794 Intron NP_001258737.1
NM_178174.3 794 Intron NP_835468.1
XM_017010822.1 794 Intron XP_016866311.1
XM_017010823.1 794 Intron XP_016866312.1
XM_017010824.1 794 Intron XP_016866313.1
XM_017010825.1 794 Intron XP_016866314.1

View Full Product Details