Product Details

SNP ID
rs199914113
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:26204650 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCGTTGGTGTGTTGGTCATGTC[G/T]GGTCGCGGCAAAGGCGGAAAGGGAC
Phenotype
MIM: 602792 MIM: 602804 MIM: 602811 MIM: 602830
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HIST1H2AD PubMed Links

Gene Details

Gene
HIST1H2AD
Gene Name
histone cluster 1, H2ad
There are no transcripts associated with this gene.

Gene
HIST1H2BF
Gene Name
histone cluster 1, H2bf
There are no transcripts associated with this gene.

Gene
HIST1H3D
Gene Name
histone cluster 1, H3d
There are no transcripts associated with this gene.

Gene
HIST1H4E
Gene Name
histone cluster 1, H4e
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003545.3 Intron NP_003536.1

View Full Product Details