Product Details

SNP ID
rs199916450
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:159907076 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCATCGTGCACTGGGTCATTATG[A/T]GCATCTCCCCAGTGGGGTTTGTTGA
Phenotype
MIM: 165180
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MAS1 PubMed Links

Gene Details

Gene
MAS1
Gene Name
MAS1 proto-oncogene, G protein-coupled receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002377.2 135 Missense Mutation AGC,TGC S41C NP_002368.1

View Full Product Details