Product Details

SNP ID
rs199541580
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28125649 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTGTGGGAGCTCTGCCGTCAGT[A/G]GCTGAGGCCAGAATGCCACACCAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZSCAN16 PubMed Links

Gene Details

Gene
ZSCAN16
Gene Name
zinc finger and SCAN domain containing 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320555.1 359 Nonsense Mutation TAG,TGG *69W NP_001307484.1
NM_001320556.1 359 Nonsense Mutation TAG,TGG *69W NP_001307485.1
NM_001320557.1 359 Nonsense Mutation TAG,TGG *69W NP_001307486.1
NM_001320558.1 359 Nonsense Mutation TAG,TGG *69W NP_001307487.1
NM_025231.2 359 Nonsense Mutation TAG,TGG *69W NP_079507.1
XM_017011324.1 359 Nonsense Mutation TAG,TGG *69W XP_016866813.1
Gene
ZSCAN16-AS1
Gene Name
ZSCAN16 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details