Product Details

SNP ID
rs199819622
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:73518196 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCAGCAAACTTGCATGCAATGTG[A/G]GCCGTGTGGCAATCCAATACAGGGG
Phenotype
MIM: 130590
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EEF1A1 PubMed Links

Gene Details

Gene
EEF1A1
Gene Name
eukaryotic translation elongation factor 1 alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001402.5 1161 Silent Mutation GCC,GCT A366A NP_001393.1
XM_011535514.2 1161 Intron XP_011533816.1

View Full Product Details