Product Details

SNP ID
rs200115914
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43517143 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCAGGCGGTGGAGGAAATGCGG[A/G]GCCGCGTGGTTCTGGGGGAGTTTGG
Phenotype
MIM: 610060 MIM: 607845 MIM: 609775
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC73 PubMed Links

Gene Details

Gene
LRRC73
Gene Name
leucine rich repeat containing 73
There are no transcripts associated with this gene.

Gene
POLR1C
Gene Name
RNA polymerase I subunit C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318876.1 120 Missense Mutation AGC,GGC S12G NP_001305805.1
NM_203290.3 120 Missense Mutation AGC,GGC S12G NP_976035.1
XM_011515000.2 120 Missense Mutation AGC,GGC S12G XP_011513302.1
Gene
XPO5
Gene Name
exportin 5
There are no transcripts associated with this gene.

Gene
YIPF3
Gene Name
Yip1 domain family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015388.3 120 Intron NP_056203.2
XM_005248989.4 120 Intron XP_005249046.1
XM_005248990.2 120 Intron XP_005249047.1
XM_011514458.2 120 Intron XP_011512760.1

View Full Product Details