Product Details
- SNP ID
-
rs199583279
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:7727593 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACAGCGCCTTCCTCAACGACGCGG[A/G]CATGGTCATGAGCTTTGTGAACCTG
- Phenotype
-
MIM: 112266
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BMP6
PubMed Links
Gene Details
- Gene
- BMP6
- Gene Name
- bone morphogenetic protein 6
There are no transcripts associated with this gene.
View Full Product Details