Product Details

SNP ID
rs199682504
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:89257709 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAATAAATTAAGTTGAAAAATAT[G/T]TAGGAGGCAGGGAATATCAACCTAG
Phenotype
MIM: 137162
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GABRR2 PubMed Links

Gene Details

Gene
GABRR2
Gene Name
gamma-aminobutyric acid type A receptor rho2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002043.4 2578 Nonsense Mutation TAA,TAC *453Y NP_002034.3
XM_011535713.2 2578 Nonsense Mutation TAA,TAC *409Y XP_011534015.1
XM_011535714.2 2578 Nonsense Mutation TAA,TAC *354Y XP_011534016.1
XM_011535715.2 2578 Nonsense Mutation TAA,TAC *354Y XP_011534017.1
XM_011535716.2 2578 Nonsense Mutation TAA,TAC *354Y XP_011534018.1
XM_011535717.2 2578 Nonsense Mutation TAA,TAC *354Y XP_011534019.1

View Full Product Details