Product Details
- SNP ID
-
rs200194994
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:28505734 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAAGTAAAGACCTTCTGTTCTTTT[A/T]CTCCATTCACATCCCCTTTCTCAAA
- Phenotype
-
MIM: 607913
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GPX6
PubMed Links
Gene Details
- Gene
- GPX6
- Gene Name
- glutathione peroxidase 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_182701.1 |
478 |
Missense Mutation |
GAA,GTA |
E143V |
NP_874360.1 |
View Full Product Details