Product Details

SNP ID
rs199526198
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:89644132 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATGGGAACTCTTCCATGTAGGAT[C/T]GGATTTCAGGCATCTCTCCAGGTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101929057 PubMed Links

Gene Details

Gene
LOC101929057
Gene Name
uncharacterized LOC101929057
There are no transcripts associated with this gene.

Gene
LYRM2
Gene Name
LYR motif containing 2
There are no transcripts associated with this gene.

Gene
MDN1
Gene Name
midasin AAA ATPase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014611.2 16656 Missense Mutation CAA,CGA Q5555R NP_055426.1
XM_005248700.3 16656 Missense Mutation CAA,CGA Q3550R XP_005248757.1
XM_006715405.3 16656 Missense Mutation CAA,CGA Q3703R XP_006715468.1
XM_011535635.2 16656 Missense Mutation CAA,CGA Q5458R XP_011533937.1
XM_011535636.2 16656 Intron XP_011533938.1

View Full Product Details