Product Details

SNP ID
rs199941858
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:132588811 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGGCTGAGTTGAAGTAAGCAAA[C/T]CAGATAAAGATGTCAAAGACCAGTG
Phenotype
MIM: 607405
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAAR5 PubMed Links

Gene Details

Gene
TAAR5
Gene Name
trace amine associated receptor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003967.2 928 Nonsense Mutation TGA,TGG *292W NP_003958.2

View Full Product Details