Product Details
- SNP ID
-
rs199942265
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:88143874 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCAGGCTCACAGAGCCTCGGCAGA[C/T]GTGTCTGTGGACACAGACATGGTTA
- Phenotype
-
MIM: 114610
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CNR1
PubMed Links
Gene Details
- Gene
- CNR1
- Gene Name
- cannabinoid receptor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001160226.1 |
1657 |
Silent Mutation |
ACA,ACG |
T467T |
NP_001153698.1 |
NM_001160258.1 |
1657 |
Silent Mutation |
ACA,ACG |
T467T |
NP_001153730.1 |
NM_001160259.1 |
1657 |
Silent Mutation |
ACA,ACG |
T467T |
NP_001153731.1 |
NM_016083.4 |
1657 |
Silent Mutation |
ACA,ACG |
T467T |
NP_057167.2 |
NM_033181.3 |
1657 |
Silent Mutation |
ACA,ACG |
T434T |
NP_149421.2 |
XM_006715330.3 |
1657 |
Silent Mutation |
ACA,ACG |
T467T |
XP_006715393.1 |
XM_011535424.1 |
1657 |
Silent Mutation |
ACA,ACG |
T467T |
XP_011533726.1 |
XM_011535425.1 |
1657 |
Silent Mutation |
ACA,ACG |
T467T |
XP_011533727.1 |
XM_011535426.1 |
1657 |
Silent Mutation |
ACA,ACG |
T467T |
XP_011533728.1 |
XM_011535428.1 |
1657 |
Silent Mutation |
ACA,ACG |
T467T |
XP_011533730.1 |
XM_017010238.1 |
1657 |
Silent Mutation |
ACA,ACG |
T467T |
XP_016865727.1 |
XM_017010239.1 |
1657 |
Silent Mutation |
ACA,ACG |
T467T |
XP_016865728.1 |
XM_017010240.1 |
1657 |
Silent Mutation |
ACA,ACG |
T467T |
XP_016865729.1 |
View Full Product Details