Product Details

SNP ID
rs200158792
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:142753500 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCTCCTGTTCCCGCTCTGTTGC[G/T]TTTAGGCTGTCTTCCGAAGTGCTCT
Phenotype
MIM: 143054
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
HIVEP2 PubMed Links

Gene Details

Gene
HIVEP2
Gene Name
human immunodeficiency virus type I enhancer binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006734.3 7656 Missense Mutation AAA,AAC K2316N NP_006725.3
XM_017010805.1 7656 Missense Mutation AAA,AAC K2316N XP_016866294.1

View Full Product Details