Product Details

SNP ID
rs199963563
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:149598294 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGTTCTTCAGAAGTCCCTCGGCG[A/G]CTACAGATGGAGTCTATCTCATCAA
Phenotype
MIM: 606696
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GINM1 PubMed Links

Gene Details

Gene
GINM1
Gene Name
glycoprotein integral membrane 1
There are no transcripts associated with this gene.

Gene
KATNA1
Gene Name
katanin catalytic subunit A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204076.1 1294 Silent Mutation AGC,AGT S239S NP_001191005.1
NM_007044.3 1294 Silent Mutation AGC,AGT S315S NP_008975.1
XM_005266804.2 1294 Silent Mutation AGC,AGT S315S XP_005266861.1
XM_005266805.3 1294 Silent Mutation AGC,AGT S262S XP_005266862.1
XM_017010207.1 1294 Silent Mutation AGC,AGT S315S XP_016865696.1
XM_017010208.1 1294 Silent Mutation AGC,AGT S187S XP_016865697.1
XM_017010209.1 1294 Silent Mutation AGC,AGT S187S XP_016865698.1
XM_017010210.1 1294 Silent Mutation AGC,AGT S315S XP_016865699.1
Gene
RPS18P9
Gene Name
ribosomal protein S18 pseudogene 9
There are no transcripts associated with this gene.

View Full Product Details