Product Details
- SNP ID
-
rs199548273
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:144095400 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAGTACGAGTCGCGGTGTTGGTTC[A/C]CCAGCCACTCCCACTTGGTGGTGTC
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SF3B5
PubMed Links
Gene Details
- Gene
- SF3B5
- Gene Name
- splicing factor 3b subunit 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_031287.2 |
218 |
Missense Mutation |
GGG,GTG |
G33V |
NP_112577.1 |
View Full Product Details