Product Details
- SNP ID
-
rs199865478
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:132570500 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGTGATGATTTCAATCCTCCATT[A/T]CAAGCAGCTGCACTCTCCGACCAAT
- Phenotype
-
MIM: 608923
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TAAR6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs8192622] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TAAR6
- Gene Name
- trace amine associated receptor 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_175067.1 |
179 |
Missense Mutation |
TAC,TTC |
Y60F |
NP_778237.1 |
View Full Product Details