Product Details

SNP ID
rs199550556
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:125986555 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGCACGGTGGGCAGCTGCAATGG[C/T]GCTGTCGTGTACCCTTAACAGGTAT
Phenotype
MIM: 609998
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HINT3 PubMed Links

Gene Details

Gene
HINT3
Gene Name
histidine triad nucleotide binding protein 3
There are no transcripts associated with this gene.

Gene
TRMT11
Gene Name
tRNA methyltransferase 11 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031712.2 27 Intron NP_001026882.2
XM_017011168.1 27 UTR 5 XP_016866657.1
XM_017011169.1 27 UTR 5 XP_016866658.1

View Full Product Details