Product Details

SNP ID
rs199907558
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:150520016 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGCTCCCTGAGGATCGTTCTGGT[A/G]GGGAAAACTGGAAGTGGGAAAAGTG
Phenotype
MIM: 616961
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GIMAP7 PubMed Links

Gene Details

Gene
GIMAP7
Gene Name
GTPase, IMAP family member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153236.3 172 Silent Mutation GTA,GTG V14V NP_694968.1

View Full Product Details