Product Details

SNP ID
rs200318770
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:40133082 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTAACAAAAGTATCTTCCTTTTT[C/T]GCCTGTGAATGTTTGAGTATTGCTG
Phenotype
MIM: 609188 MIM: 609187
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MPLKIP PubMed Links

Gene Details

Gene
MPLKIP
Gene Name
M-phase specific PLK1 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138701.3 602 Missense Mutation NP_619646.1
Gene
SUGCT
Gene Name
succinyl-CoA:glutarate-CoA transferase
There are no transcripts associated with this gene.

View Full Product Details