Product Details

SNP ID
rs200699755
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:142751966 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCACCATCTCTCTGCCCACCGC[C/T]CCTCCAGCCACTGGCACGAAGTGCC
Phenotype
MIM: 276000
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRSS1 PubMed Links

Gene Details

Gene
PRSS1
Gene Name
protease, serine 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002769.4 Intron NP_002760.1
XM_011516411.1 Intron XP_011514713.1

View Full Product Details