Product Details

SNP ID
rs200887444
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:155071065 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTTTCTGGTGGCGGCGACGTTC[G/T]CCTGGAACCTGCTGGTGCTGGCGAC
Phenotype
MIM: 601305
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HTR5A PubMed Links

Gene Details

Gene
HTR5A
Gene Name
5-hydroxytryptamine receptor 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024012.3 742 Missense Mutation GCC,TCC A56S NP_076917.1
Gene
HTR5A-AS1
Gene Name
HTR5A antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details